Assay Details
| Gene Symbol: PEBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198642.1 | NP_001185571.1 | 1 - 2 | 193 | 68 | |
| GenBank mRNA | BX933434.2 | - | 1 - 2 | 154 | 68 | |
| BX934603.1 | - | 1 - 2 | 177 | 68 | ||
| CN222635.1 | - | 1 - 2 | 193 | 68 | ||
More Information
Set Membership: |
|

PEBP1
Genomic Map