Assay Details
| Gene Symbol: GUCA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197038.1 | NP_001183967.1 | 1 - 2 | 183 | 106 | |
| GenBank mRNA | BX934866.1 | - | 1 - 2 | 147 | 106 | |
| BX934917.2 | - | 1 - 2 | 177 | 106 | ||
More Information
Set Membership: |
|

GUCA2B
Genomic Map