Assay Details
| Gene Symbol: EIF3I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164395.1 | NP_001157867.1 | 7 - 8 | 693 | 70 | |
| GenBank mRNA | CR352485.1 | - | 6 - 7 | 770 | 70 | |
| CR385826.1 | - | 2 - 3 | 201 | 70 | ||
More Information
Set Membership: |
|

EIF3I
Genomic Map