Assay Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025154769.1 | XP_025010537.1 | 6 - 7 | 896 | 111 | |
| XM_414345.6 | XP_414345.3 | 7 - 8 | 938 | 111 | ||
| GenBank mRNA | CR387191.1 | - | 1 - 2 | 92 | 111 | |
More Information
Set Membership: |
|

ASB14
Genomic Map