Assay Details
| Gene Symbol: PDK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199909.1 | NP_001186838.1 | 9 - 10 | 1083 | 62 | |
| GenBank mRNA | CR387492.1 | - | 9 - 10 | 1079 | 62 | |
More Information
Set Membership: |
|

PDK4
Genomic Map