Assay Details
| Gene Symbol: COL8A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293134.1 | NP_001280063.1 | 5 | 3976 | 85 | |
| XM_004938241.3 | XP_004938298.1 | 4 | 3968 | 85 | ||
| XM_015296025.2 | XP_015151511.1 | 6 | 4164 | 85 | ||
| GenBank mRNA | CR387830.1 | - | 1 | 1453 | 85 | |
More Information
Set Membership: |
|

COL8A1
Genomic Map