Assay Details
| Gene Symbol: PAPOLB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198601.3 | NP_001185530.1 | 11 - 12 | 1303 | 78 | |
| XM_015287288.2 | XP_015142774.1 | 11 - 12 | 1310 | 78 | ||
| XM_015287290.2 | XP_015142776.1 | 11 - 12 | 1310 | 78 | ||
| XM_015287291.2 | XP_015142777.1 | 11 - 12 | 1310 | 78 | ||
| GenBank mRNA | AF061758.1 | - | 7 - 8 | 676 | 78 | |
More Information
Set Membership: |
|

PAPOLB
Genomic Map