Assay Details
| Gene Symbol: PSMD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292690.2 | XP_015148176.1 | 6 - 7 | 598 | 58 | |
| GenBank mRNA | AJ719835.1 | - | 6 - 7 | 598 | 58 | |
More Information
Set Membership: |
|

PSMD7
Genomic Map