Assay Details
| Gene Symbol: AQP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001231780.5 | XP_001231781.2 | 1 - 2 | 460 | 96 | |
| GenBank mRNA | AJ829443.3 | - | 1 - 2 | 304 | 96 | |
More Information
Set Membership: |
|

AQP5
Genomic Map