Assay Details
| Gene Symbol: FAM21C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012593.2 | NP_001012611.2 | 28 - 29 | 3093 | 68 | |
| XM_015288386.2 | XP_015143872.1 | 28 - 29 | 3169 | 68 | ||
| XM_015288389.2 | XP_015143875.1 | 27 - 28 | 3106 | 68 | ||
| XM_015288390.2 | XP_015143876.1 | 27 - 28 | 3100 | 68 | ||
| XM_025151585.1 | XP_025007353.1 | 28 - 29 | 3184 | 68 | ||
| XM_025151586.1 | XP_025007354.1 | 28 - 29 | 3181 | 68 | ||
| XM_025151587.1 | XP_025007355.1 | 28 - 29 | 3178 | 68 | ||
| XM_025151588.1 | XP_025007356.1 | 27 - 28 | 3127 | 68 | ||
| XM_025151589.1 | XP_025007357.1 | 27 - 28 | 3010 | 68 | ||
| GenBank mRNA | AJ851808.1 | - | 28 - 29 | 3095 | 68 | |
More Information
Set Membership: |
|

FAM21C
Genomic Map