Assay Details
| Gene Symbol: GGT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934343.3 | XP_004934400.2 | 6 - 7 | 911 | 66 | |
| XM_015275447.2 | XP_015130933.1 | 6 - 7 | 871 | 66 | ||
| XM_015275448.2 | XP_015130934.1 | 6 - 7 | 818 | 66 | ||
| XM_015275449.2 | XP_015130935.1 | 6 - 7 | 871 | 66 | ||
| XM_015275450.2 | XP_015130936.1 | 6 - 7 | 862 | 66 | ||
| XM_015275451.2 | XP_015130937.1 | 6 - 7 | 910 | 66 | ||
| XM_015275452.2 | XP_015130938.1 | 6 - 7 | 896 | 66 | ||
| XM_015275453.2 | XP_015130939.1 | 6 - 7 | 1332 | 66 | ||
| XM_025155514.1 | XP_025011282.1 | 6 - 7 | 758 | 66 | ||
| XM_025155515.1 | XP_025011283.1 | 6 - 7 | 861 | 66 | ||
| XM_025155516.1 | XP_025011284.1 | 6 - 7 | 819 | 66 | ||
| GenBank mRNA | AM050164.1 | - | 4 - 5 | 501 | 66 | |
More Information
Set Membership: |
|

GGT2
Genomic Map