Assay Details
Gene Symbol: CHST15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_421811.6 | XP_421811.2 | 2 - 3 | 1156 | 121 | |
GenBank mRNA | BX929349.1 | - | 2 - 3 | 1152 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
