Assay Details
Gene Symbol: FILIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284757.2 | XP_015140243.1 | 3 - 4 | 898 | 101 | |
XM_015284758.2 | XP_015140244.1 | 4 - 5 | 1040 | 101 | ||
XM_015284759.2 | XP_015140245.1 | 4 - 5 | 1036 | 101 | ||
XM_015284761.2 | XP_015140247.1 | 4 - 5 | 1045 | 101 | ||
XM_419877.6 | XP_419877.5 | 4 - 5 | 1039 | 101 | ||
GenBank mRNA | BX929400.1 | - | 3 - 4 | 649 | 101 | |
BX935837.2 | - | 4 - 5 | 804 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
