Assay Details
| Gene Symbol: PITX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167684.1 | NP_001161156.1 | 2 - 3 | 658 | 79 | |
| NM_001167685.1 | NP_001161157.1 | 3 - 4 | 405 | 79 | ||
| NM_001167686.1 | NP_001161158.1 | 3 - 4 | 502 | 79 | ||
| XM_015293986.2 | XP_015149472.2 | 3 - 4 | 674 | 79 | ||
| GenBank mRNA | AF069397.1 | - | 2 - 3 | 366 | 79 | |
| BX929872.1 | - | 2 - 3 | 657 | 79 | ||
| BX930965.1 | - | 1 - 2 | 620 | 79 | ||
| CD763774.1 | - | 3 - 4 | 515 | 79 | ||
More Information
Set Membership: |
|

PITX1
Genomic Map