Assay Details
Gene Symbol: QSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199414.1 | NP_001186343.1 | 2 - 3 | 244 | 64 | |
XM_015286155.2 | XP_015141641.2 | 3 - 4 | 766 | 64 | ||
XM_015286156.2 | XP_015141642.1 | 3 - 4 | 507 | 64 | ||
GenBank mRNA | CR389945.1 | - | 2 - 3 | 250 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
