Assay Details
| Gene Symbol: C8H1orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277560.1 | NP_001264489.1 | 2 - 3 | 472 | 98 | |
| GenBank mRNA | BU268876.1 | - | 1 - 2 | 74 | 98 | |
| CR390338.1 | - | 2 - 3 | 463 | 98 | ||
More Information
Set Membership: |
|

C8H1orf52
Genomic Map