Assay Details
Gene Symbol: SLC16A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004942143.3 | XP_004942200.1 | 4 - 5 | 860 | 89 | |
XM_015288412.2 | XP_015143898.1 | 4 - 5 | 1099 | 89 | ||
XM_025152015.1 | XP_025007783.1 | 5 - 6 | 771 | 89 | ||
XM_025152016.1 | XP_025007784.1 | 4 - 5 | 574 | 89 | ||
XM_025152017.1 | XP_025007785.1 | 4 - 5 | 620 | 89 | ||
XM_025152018.1 | XP_025007786.1 | 4 - 5 | 796 | 89 | ||
XM_421663.6 | XP_421663.3 | 4 - 5 | 1275 | 89 | ||
GenBank mRNA | CR391366.1 | - | 1 - 2 | 129 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
