Assay Details
| Gene Symbol: ACOT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282414.2 | XP_015137900.1 | 1 - 2 | 148 | 94 | |
| GenBank mRNA | BX929282.2 | - | 1 - 2 | 103 | 94 | |
| BX932127.1 | - | 1 - 2 | 109 | 94 | ||
| BX932290.1 | - | 1 - 2 | 136 | 94 | ||
| BX935853.1 | - | 1 - 2 | 112 | 94 | ||
| CR406050.1 | - | 1 - 2 | 114 | 94 | ||
More Information
Set Membership: |
|

ACOT13
Genomic Map