Assay Details
| Gene Symbol: FABP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199476.1 | NP_001186405.1 | 2 - 3 | 317 | 98 | |
| GenBank mRNA | BX935863.1 | - | 2 - 3 | 300 | 98 | |
| CR523331.1 | - | 2 - 3 | 317 | 98 | ||
More Information
Set Membership: |
|

FABP9
Genomic Map