Assay Details
Gene Symbol: TNX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015294997.2 | XP_015150483.2 | 3 - 4 | 581 | 124 | |
XM_025155795.1 | XP_025011563.1 | 3 - 4 | 581 | 124 | ||
XM_025155796.1 | XP_025011564.1 | 3 - 4 | 581 | 124 | ||
XM_025155797.1 | XP_025011565.1 | 3 - 4 | 581 | 124 | ||
XM_025155798.1 | XP_025011566.1 | 3 - 4 | 581 | 124 | ||
XM_025155800.1 | XP_025011568.1 | 3 - 4 | 581 | 124 | ||
XM_025155801.1 | XP_025011569.1 | 3 - 4 | 581 | 124 | ||
XM_025155802.1 | XP_025011570.1 | 3 - 4 | 581 | 124 | ||
XM_025155803.1 | XP_025011571.1 | 3 - 4 | 581 | 124 | ||
XM_025155804.1 | XP_025011572.1 | 3 - 4 | 581 | 124 | ||
XM_025155805.1 | XP_025011573.1 | 3 - 4 | 581 | 124 | ||
XM_025155806.1 | XP_025011574.1 | 3 - 4 | 581 | 124 | ||
XM_025155807.1 | XP_025011575.1 | 3 - 4 | 581 | 124 | ||
XM_025155808.1 | XP_025011576.1 | 3 - 4 | 581 | 124 | ||
XM_025155809.1 | XP_025011577.1 | 3 - 4 | 581 | 124 | ||
XM_025155810.1 | XP_025011578.1 | 3 - 4 | 581 | 124 | ||
XM_025155811.1 | XP_025011579.1 | 3 - 4 | 581 | 124 | ||
GenBank mRNA | X99062.1 | - | 974 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
