Assay Details
| Gene Symbol: FKBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418735.6 | XP_418735.1 | 1 - 2 | 370 | 58 | |
| GenBank mRNA | BX930269.1 | - | 1 - 2 | 289 | 58 | |
More Information
Set Membership: |
|

FKBP14
Genomic Map