Assay Details
Gene Symbol: IMMP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277356.1 | NP_001264285.1 | 2 - 3 | 101 | ||
NM_001277357.1 | NP_001264286.1 | 2 - 3 | 101 | |||
XM_025147649.1 | XP_025003417.1 | 2 - 3 | 101 | |||
XM_025147655.1 | XP_025003423.1 | 2 - 3 | 101 | |||
XM_025147656.1 | XP_025003424.1 | 2 - 3 | 101 | |||
XM_025147657.1 | XP_025003425.1 | 2 - 3 | 101 | |||
XM_025147659.1 | XP_025003427.1 | 2 - 3 | 101 | |||
XM_025147660.1 | XP_025003428.1 | 2 - 3 | 101 | |||
GenBank mRNA | BX930706.1 | - | 2 - 3 | 225 | 101 | |
BX931538.1 | - | 2 - 3 | 101 | |||
CF253209.1 | - | 2 - 3 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
