Assay Details
| Gene Symbol: FUNDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420194.6 | XP_420194.1 | 3 - 4 | 287 | 92 | |
| GenBank mRNA | BX930592.1 | - | 3 - 4 | 277 | 92 | |
| BX931014.1 | - | 3 - 4 | 407 | 92 | ||
| CR406059.1 | - | 3 - 4 | 280 | 92 | ||
More Information
Set Membership: |
|

FUNDC2
Genomic Map