Assay Details
| Gene Symbol: LOC416147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025155025.1 | XP_025010793.1 | 6 - 7 | 3208 | 75 | |
| XM_025155026.1 | XP_025010794.1 | 6 - 7 | 3208 | 75 | ||
| XM_414478.6 | XP_414478.5 | 6 - 7 | 3206 | 75 | ||
| GenBank mRNA | BX931153.1 | - | 1 - 2 | 605 | 75 | |
| BX950599.1 | - | 1 - 2 | 667 | 75 | ||
| MG066609.1 | - | 1 - 2 | 636 | 75 | ||
| MG066616.1 | - | 1 - 2 | 636 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025155025.1, XM_025155026.1 transcripts.
Set Membership: |
|

LOC416147
Genomic Map