Assay Details
| Gene Symbol: PRORSD1P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195561.1 | NP_001182490.1 | 1 - 2 | 103 | 101 | |
| GenBank mRNA | BX929534.2 | - | 1 - 2 | 108 | 101 | |
| BX931870.1 | - | 1 - 2 | 162 | 101 | ||
| BX936017.1 | - | 1 - 2 | 186 | 101 | ||
More Information
Set Membership: |
|

PRORSD1P
Genomic Map