Assay Details
Gene Symbol: REEP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199532.1 | NP_001186461.1 | 4 - 5 | 370 | 78 | |
XM_015288099.1 | XP_015143585.1 | 4 - 5 | 348 | 78 | ||
XM_025151515.1 | XP_025007283.1 | 3 - 4 | 280 | 78 | ||
GenBank mRNA | BX932673.1 | - | 4 - 5 | 370 | 78 | |
CR387172.1 | - | 4 - 5 | 534 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
