Assay Details
| Gene Symbol: OLAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281988.2 | XP_015137474.1 | 3 - 4 | 2752 | 89 | |
| XM_015281989.2 | XP_015137475.1 | 2 - 3 | 2634 | 89 | ||
| XM_418634.5 | XP_418634.1 | 2 - 3 | 2618 | 89 | ||
| GenBank mRNA | BX934632.1 | - | 2 - 3 | 216 | 89 | |
More Information
Set Membership: |
|

OLAH
Genomic Map