Assay Details
Gene Symbol: RFX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291868.2 | XP_015147354.2 | 9 | 3659 | 61 | |
XM_025154100.1 | XP_025009868.1 | 8 | 4415 | 61 | ||
XM_429081.6 | XP_429081.6 | 12 | 3426 | 61 | ||
GenBank mRNA | BX934980.1 | - | 1 | 1548 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
