Assay Details
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001232398.5 | XP_001232399.1 | 5 - 6 | 473 | 76 | |
XM_001232443.5 | XP_001232444.1 | 4 - 5 | 474 | 76 | ||
XM_004944551.3 | XP_004944608.1 | 5 - 6 | 563 | 76 | ||
XM_004944552.3 | XP_004944609.1 | 4 - 5 | 503 | 76 | ||
XM_015293413.2 | XP_015148899.1 | 5 - 6 | 582 | 76 | ||
XM_015293414.2 | XP_015148900.1 | 5 - 6 | 455 | 76 | ||
XM_015293415.2 | XP_015148901.1 | 5 - 6 | 643 | 76 | ||
XM_414329.5 | XP_414329.1 | 5 - 6 | 545 | 76 | ||
GenBank mRNA | BX935395.1 | - | 5 - 6 | 472 | 76 | |
CR353336.1 | - | 3 - 4 | 835 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
