Assay Details
| Gene Symbol: NDUFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302167.1 | NP_001289096.1 | 1 - 2 | 284 | 144 | |
| GenBank mRNA | BX929464.1 | - | 1 - 2 | 175 | 150 | |
| BX935929.1 | - | 1 - 2 | 281 | 150 | ||
| CR338759.1 | - | 1 - 2 | 284 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302167.1 transcripts.
Set Membership: |
|

NDUFB3
Genomic Map