Assay Details
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015293332.2 | XP_015148818.1 | 5 - 6 | 1004 | 80 | |
GenBank mRNA | BX936097.1 | - | 2 - 3 | 346 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
