Assay Details
| Gene Symbol: PLA2G2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277914.1 | NP_001264843.1 | 2 - 3 | 216 | 102 | |
| GenBank mRNA | BU270619.1 | - | 2 - 3 | 196 | 102 | |
| BX950757.1 | - | 2 - 3 | 216 | 102 | ||
More Information
Set Membership: |
|

PLA2G2A
Genomic Map