Assay Details
Gene Symbol: CMTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414155.6 | XP_414155.4 | 3 - 4 | 12400 | 84 | |
GenBank mRNA | CR338861.1 | - | 2 - 3 | 233 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
