Assay Details
| Gene Symbol: NENF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004935332.3 | XP_004935389.1 | 1 - 2 | 566 | 63 | |
| XM_419430.5 | XP_419430.1 | 1 - 2 | 163 | 63 | ||
| GenBank mRNA | CR353202.1 | - | 1 - 2 | 152 | 63 | |
More Information
Set Membership: |
|

NENF
Genomic Map