Assay Details
Gene Symbol: CST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199394.1 | NP_001186323.1 | 3 - 4 | 583 | 92 | |
GenBank mRNA | AM070770.1 | - | 3 - 4 | 586 | 92 | |
BX932453.1 | - | 3 - 4 | 583 | 92 | ||
BX932714.1 | - | 3 - 4 | 369 | 92 | ||
BX932920.2 | - | 3 - 4 | 417 | 92 | ||
CR353763.1 | - | 3 - 4 | 413 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
