Assay Details
Gene Symbol: LOC107057437 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015302512.2 | XP_015157998.2 | 8 - 9 | 2123 | 67 | |
XM_015302515.2 | XP_015158001.2 | 7 - 8 | 2015 | 67 | ||
XM_025143244.1 | XP_024999012.1 | 9 - 10 | 2455 | 67 | ||
XM_025143245.1 | XP_024999013.1 | 12 - 13 | 4239 | 67 | ||
XM_025143246.1 | XP_024999014.1 | 12 - 13 | 5635 | 67 | ||
XM_025143247.1 | XP_024999015.1 | 10 - 11 | 2366 | 67 | ||
XM_025143248.1 | XP_024999016.1 | 8 - 9 | 2793 | 67 | ||
XM_025143249.1 | XP_024999017.1 | 9 - 10 | 2288 | 67 | ||
XM_025143250.1 | XP_024999018.1 | 10 - 11 | 2636 | 67 | ||
XM_025143251.1 | XP_024999019.1 | 10 - 11 | 2588 | 67 | ||
XM_025143252.1 | XP_024999020.1 | 9 - 10 | 1622 | 67 | ||
XM_025143253.1 | XP_024999021.1 | 8 - 9 | 763 | 67 | ||
GenBank mRNA | CR354154.1 | - | 1 - 2 | 306 | 67 |
Gene Symbol: RIMBP3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025143262.1 | XP_024999030.1 | 7 - 8 | 1189 | 67 | |
XM_025143263.1 | XP_024999031.1 | 6 - 7 | 1081 | 67 | ||
XM_025143264.1 | XP_024999032.1 | 10 - 11 | 4800 | 67 | ||
XM_025143265.1 | XP_024999033.1 | 9 - 10 | 2761 | 67 | ||
XM_025143266.1 | XP_024999034.1 | 10 - 11 | 5003 | 67 | ||
XM_025143267.1 | XP_024999035.1 | 10 - 11 | 4742 | 67 | ||
XM_025143268.1 | XP_024999036.1 | 10 - 11 | 5387 | 67 | ||
XM_025143269.1 | XP_024999037.1 | 10 - 11 | 3654 | 67 | ||
XM_025143270.1 | XP_024999038.1 | 7 - 8 | 1770 | 67 | ||
XM_025143271.1 | XP_024999039.1 | 8 - 9 | 813 | 67 | ||
XM_025143272.1 | XP_024999040.1 | 8 - 9 | 5533 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
