Assay Details
Gene Symbol: C1GALT1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277789.1 | NP_001264718.1 | 1 - 2 | 53 | 74 | |
GenBank mRNA | BX930232.1 | - | 1 - 2 | 48 | 74 | |
BX931310.2 | - | 1 - 2 | 53 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001277789.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
