Assay Details
Gene Symbol: SYNGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252278.3 | NP_001239207.3 | 2 - 3 | 66 | ||
XM_015285214.2 | XP_015140700.2 | 2 - 3 | 66 | |||
XM_015285219.2 | XP_015140705.1 | 2 - 3 | 66 | |||
GenBank mRNA | BX931567.2 | - | 2 - 3 | 417 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
