Assay Details
| Gene Symbol: GIPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422385.6 | XP_422385.1 | 2 - 3 | 543 | 69 | |
| GenBank mRNA | BX932652.2 | - | 2 - 3 | 467 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GIPC2| Gene Symbol: GIPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422385.6 | XP_422385.1 | 2 - 3 | 543 | 69 | |
| GenBank mRNA | BX932652.2 | - | 2 - 3 | 467 | 69 | |
Set Membership: |
|