Assay Details
| Gene Symbol: CHAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199656.1 | NP_001186585.1 | 2 - 3 | 284 | 125 | |
| GenBank mRNA | BX933277.2 | - | 2 - 3 | 267 | 125 | |
| CR353343.1 | - | 2 - 3 | 287 | 125 | ||
More Information
Set Membership: |
|

CHAC1
Genomic Map