Assay Details
| Gene Symbol: ARPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277486.1 | NP_001264415.1 | 5 - 6 | 439 | 62 | |
| GenBank mRNA | BX933901.2 | - | 5 - 6 | 508 | 62 | |
| BX934934.1 | - | 5 - 6 | 449 | 62 | ||
| CN220727.1 | - | 5 - 6 | 439 | 62 | ||
More Information
Set Membership: |
|

ARPC3
Genomic Map