Assay Details
Gene Symbol: AKIRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193595.2 | NP_001180524.1 | 1 | 109 | 149 | |
GenBank mRNA | BX934526.2 | - | 1 | 109 | 149 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193595.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
