Assay Details
Gene Symbol: AKIRIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193595.2 | NP_001180524.1 | 4 - 5 | 652 | 66 | |
GenBank mRNA | BU128275.1 | - | 3 - 4 | 311 | 66 | |
BX934526.2 | - | 4 - 5 | 652 | 66 | ||
BX935737.2 | - | 5 - 6 | 514 | 66 | ||
CR387510.1 | - | 4 - 5 | 625 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
