Assay Details
| Gene Symbol: C1QA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417654.6 | XP_417654.2 | 1 - 2 | 84 | 114 | |
| GenBank mRNA | BX934534.2 | - | 1 - 2 | 50 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_417654.6 transcripts.
Set Membership: |
|

C1QA
Genomic Map