Assay Details
| Gene Symbol: MAPK1IP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936482.2 | XP_004936539.2 | 2 - 3 | 1170 | 69 | |
| XM_430071.5 | XP_430071.2 | 3 - 4 | 859 | 69 | ||
| GenBank mRNA | CR352945.1 | - | 1 - 2 | 624 | 69 | |
More Information
Set Membership: |
|

MAPK1IP1L
Genomic Map