Assay Details
| Gene Symbol: SBSPONL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277502.1 | NP_001264431.1 | 1 - 2 | 203 | 123 | |
| XM_025155473.1 | XP_025011241.1 | 1 - 2 | 705 | 123 | ||
| GenBank mRNA | CR353228.1 | - | 1 - 2 | 184 | 123 | |
More Information
Set Membership: |
|

SBSPONL
Genomic Map