Assay Details
| Gene Symbol: SBSPONL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277502.1 | NP_001264431.1 | 4 - 5 | 643 | 78 | |
| GenBank mRNA | CR353228.1 | - | 4 - 5 | 624 | 78 | |
More Information
Set Membership: |
|

SBSPONL
Genomic Map