Assay Details
Gene Symbol: LSM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302170.1 | NP_001289099.1 | 2 - 3 | 150 | 113 | |
NM_001302171.1 | NP_001289100.1 | 2 - 3 | 243 | 113 | ||
GenBank mRNA | CR354370.1 | - | 2 - 3 | 150 | 113 | |
CR387219.1 | - | 2 - 3 | 243 | 113 | ||
CR391062.1 | - | 2 - 3 | 271 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302171.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
