Assay Details
| Gene Symbol: LSM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001302170.1 | NP_001289099.1 | 3 - 4 | 169 | 141 | |
| NM_001302171.1 | NP_001289100.1 | 3 - 4 | 262 | 141 | ||
| GenBank mRNA | CR354370.1 | - | 3 - 4 | 169 | 141 | |
| CR387219.1 | - | 3 - 4 | 262 | 141 | ||
| CR391062.1 | - | 3 - 4 | 290 | 141 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001302171.1 transcripts.
Set Membership: |
|

LSM5
Genomic Map