Assay Details
Gene Symbol: MALL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199428.1 | NP_001186357.1 | 1 - 2 | 444 | 68 | |
GenBank mRNA | BU298453.1 | - | 1 - 2 | 129 | 68 | |
BX929438.2 | - | 1 - 2 | 445 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
